MXT®-500 SimDist Column (Siltek®-treated stainless steel), Restek
Supplier: Restek
70104-065 70104
10851-196EA 538.16 USD
10851-196 10851-426
MXT®-500 SimDist Column (Siltek®-treated stainless steel), Restek
Chromatography Columns
Manufactured from Siltek®-treated stainless steel tubing, MXT® columns exhibit excellent peak shape and low bleed.
  • Application-specific columns in unbreakable Siltek®-treated stainless steel tubing meet all resolution criteria for high temperature simulated distillation
  • Stable to 430 °C
  • 4.5" standard coil diameter
  • Nonpolar phase
The unique MXT®-1HT SimDist methyl silicone polymer gives the correct retention time/boiling point curve. The MXT®-500 SimDist carborane siloxane polymer offers a slight shift in the calculated boiling range distribution for petroleum samples containing aromatic hydrocarbons.