Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fused Silica Tubing (Undeactivated, Untreated), Restek

Supplier: RESTEK CORP
10060 10062 10051 10050 10061 10052 10055 10057 10056
Fused silica tubing is tested for superior strength through an undeactivated, untreated delivery system.

  • Strength-tested
  • Inexpensive
  • Coiled in 7-inch diameters, banded in 5-meter sections, and mounted in a Restek cage

This economical tubing comes in 5-meter lengths that are coiled to be 7 inches in diameter.




 

ORDER

Expand All / Collapse All
Description Column Format Phase I.D. x L O.D. Packing Material Supplier No. VWR Catalog Number Unit Price Quantity
Untreated Fused Silica Tubing Capillary Silica (Si) 0.25 mm x 60 m 0.37 ± 0.04 mm Mm 0tee 10052 RK10052 Each Retrieving
Untreated Fused Silica Tubing Capillary Silica (Si) 0.25 mm x 15 m 0.37 ± 0.04 mm Mm 0tee 10050 RK10050 Each Retrieving
Untreated Fused Silica Tubing Capillary Silica (Si) 0.53 mm x 60 m 0.69 ± 0.05 mm Mm 0tee 10062 RK10062 Each Retrieving
Untreated Fused Silica Tubing Capillary Silica (Si) 0.32 mm x 30 m 0.45 ± 0.04 mm Mm 0tee 10056 RK10056 Each Retrieving
Untreated Fused Silica Tubing Capillary Silica (Si) 0.32 mm x 15 m 0.45 ± 0.04 mm Mm 0tee 10055 RK10055 Each Retrieving
Untreated Fused Silica Tubing Capillary Silica (Si) 0.53 mm x 30 m 0.69 ± 0.05 mm Mm 0tee 10061 RK10061 Each Retrieving
Untreated Fused Silica Tubing Capillary Silica (Si) 0.25 mm x 30 m 0.37 ± 0.04 mm Mm 0tee 10051 RK10051 Each Retrieving
Untreated Fused Silica Tubing Capillary Silica (Si) 0.53 mm x 15 m 0.69 ± 0.05 mm Mm 0tee 10060 RK10060 Each Retrieving
Untreated Fused Silica Tubing Capillary Silica (Si) 0.32 mm x 60 m 0.45 ± 0.04 mm Mm 0tee 10057 RK10057 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty