Anti-HLA Class II (DR) Mouse Monoclonal Antibody [clone: B-C10]

Supplier: Cell Sciences
CDM346
102995-896EA 553.85 USD
102995-896
Anti-HLA Class II (DR) Mouse Monoclonal Antibody [clone: B-C10]
Antibodies
Type: Primary
Antigen: HLA Class II (DR)
Clonality: Monoclonal
Clone: B-C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR