Anti-HLA Class II (DQ) Mouse Monoclonal Antibody [clone: B-K27]

Supplier: Cell Sciences
CDM344
102995-892EA 553.85 USD
102995-892
Anti-HLA Class II (DQ) Mouse Monoclonal Antibody [clone: B-K27]
Antibodies
Type: Primary
Antigen: HLA Class II (DQ)
Clonality: Monoclonal
Clone: B-K27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR