Anti-Biotin Mouse Monoclonal Antibody [clone: B-E24]

Supplier: Cell Sciences
CDM343
102995-890EA 526.15 USD
102995-890
Anti-Biotin Mouse Monoclonal Antibody [clone: B-E24]
Antibodies
Type: Primary
Antigen: Biotin
Clonality: Monoclonal
Clone: B-E24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR