Anti-NTSR1 Mouse Monoclonal Antibody [clone: B-N6]

Supplier: Cell Sciences
CDM342
102995-888EA 443.08 USD
102995-888
Anti-NTSR1 Mouse Monoclonal Antibody [clone: B-N6]
Antibodies
Flow Cytometry: Use 10 µl to label 10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: NTSR1
Clonality: Monoclonal
Clone: B-N6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR