Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KLRD1 Mouse Monoclonal Antibody [clone: B-D49]

Supplier: Cell Sciences
CDM332
102995-874
Anti-KLRD1 Mouse Monoclonal Antibody [clone: B-D49]
Flow Cytometry: Use 10 µl to label 10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: KLRD1
Clonality: Monoclonal
Clone: B-D49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty