Anti-GMCSF1 Mouse Monoclonal Antibody [clone: B-S37]

Supplier: Cell Sciences
CDM330
102995-870EA 553.85 USD
102995-870
Anti-GMCSF1 Mouse Monoclonal Antibody [clone: B-S37]
Antibodies
Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: B-S37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR