Anti-IL24 Mouse Monoclonal Antibody [clone: B-C54]

Supplier: Cell Sciences
CDM328
102995-866EA 955.38 USD
102995-866
Anti-IL24 Mouse Monoclonal Antibody [clone: B-C54]
Antibodies
Type: Primary
Antigen: IL24
Clonality: Monoclonal
Clone: B-C54
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR