Anti-NKP30 Mouse Monoclonal Antibody [clone: B-S30]

Supplier: Cell Sciences
CDM325
102995-860EA 443.08 USD
102995-860
Anti-NKP30 Mouse Monoclonal Antibody [clone: B-S30]
Antibodies
Flow Cytometry: Use 10 µl to label 10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: NKP30
Clonality: Monoclonal
Clone: B-S30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR