Anti-GNLY Mouse Monoclonal Antibody [clone: B-R32]

Supplier: Cell Sciences
CDM316
102995-842EA 553.85 USD
102995-842
Anti-GNLY Mouse Monoclonal Antibody [clone: B-R32]
Antibodies
Recognizes the soluble antigen

Type: Primary
Antigen: GNLY
Clonality: Monoclonal
Clone: B-R32
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR