Anti-GNLY Mouse Monoclonal Antibody [clone: B-L38]

Supplier: Cell Sciences
CDM313
102995-836EA 553.85 USD
102995-836
Anti-GNLY Mouse Monoclonal Antibody [clone: B-L38]
Antibodies
Recognizes the soluble and membrane antigen

Type: Primary
Antigen: GNLY
Clonality: Monoclonal
Clone: B-L38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR