Anti-MICB Mouse Monoclonal Antibody [clone: B-D55]

Supplier: Cell Sciences
CDM312
102995-834EA 553.85 USD
102995-834
Anti-MICB Mouse Monoclonal Antibody [clone: B-D55]
Antibodies
Recognizes the soluble antigen

Type: Primary
Antigen: MICB
Clonality: Monoclonal
Clone: B-D55
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR