Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NKP46 Mouse Monoclonal Antibody [clone: B-L46]

Supplier: Cell Sciences
CDM306
102995-824EA 540 USD InStock
102995-824
Anti-NKP46 Mouse Monoclonal Antibody [clone: B-L46]
Antibodies
Suitable for biological assay. The biological activity is measured by the ability of the coated antibody to induce degranulation of activated NK cells and to enhance IFN-g secretion by non-activated NK.

Type: Primary
Antigen: NKP46
Clonality: Monoclonal
Clone: B-L46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty