Anti-NKP46 Mouse Monoclonal Antibody [clone: B-N40]

Supplier: Cell Sciences
CDM299
102995-810EA 443.08 USD
102995-810
Anti-NKP46 Mouse Monoclonal Antibody [clone: B-N40]
Antibodies
Flow Cytometry: Use 10 µl to label 10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: NKP46
Clonality: Monoclonal
Clone: B-N40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR