Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NKP46 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-N40]

Supplier: Cell Sciences
CDM298
102995-808EA 540 USD InStock
102995-808
Anti-NKP46 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-N40]
Antibodies
Flow Cytometry: Use 10 µl to label 10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: NKP46
Clonality: Monoclonal
Clone: B-N40
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty