Anti-CXCL10/IP10 Mouse Monoclonal Antibody [clone: B-C55]

Supplier: Cell Sciences
CDM297
102995-806EA 955.38 USD
102995-806
Anti-CXCL10/IP10 Mouse Monoclonal Antibody [clone: B-C55]
Antibodies
Recognizes the soluble antigen

Type: Primary
Antigen: CXCL10/IP10
Clonality: Monoclonal
Clone: B-C55
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR