Anti-MICA Mouse Monoclonal Antibody [clone: B-N31]

Supplier: Cell Sciences
CDM290
102995-794EA 553.85 USD
102995-794
Anti-MICA Mouse Monoclonal Antibody [clone: B-N31]
Antibodies
Recognizes the soluble antigen

Type: Primary
Antigen: MICA
Clonality: Monoclonal
Clone: B-N31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR