Anti-IL27 Mouse Monoclonal Antibody [clone: B-G49]

Supplier: Cell Sciences
CDM283
102995-780EA 955.38 USD
102995-780
Anti-IL27 Mouse Monoclonal Antibody [clone: B-G49]
Antibodies
Suitable for flow cytometry on dendritic cells.

Type: Primary
Antigen: IL27
Clonality: Monoclonal
Clone: B-G49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR