Anti-IL-12 P40/p70 Mouse Monoclonal Antibody [clone: B-P40]

Supplier: Cell Sciences
CDM281
102995-776EA 955.38 USD
102995-776
Anti-IL-12 P40/p70 Mouse Monoclonal Antibody [clone: B-P40]
Antibodies
No effect on IL-12 induced proliferation on PHA and IL-2 activated lymphocytes. Suitable for intra-cytoplasmic flow cytometry.

Type: Primary
Antigen: IL-12 P40/p70
Clonality: Monoclonal
Clone: B-P40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR