Anti-IL4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-G28]

Supplier: Cell Sciences
CDM280
102995-774EA 706.15 USD
102995-774
Anti-IL4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-G28]
Antibodies
Suitable for intra-cytoplasmic flow cytometry

Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: B-G28
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR