Anti-IL4 Mouse Monoclonal Antibody [clone: B-G28]

Supplier: Cell Sciences
CDM279
102995-772EA 955.38 USD
102995-772
Anti-IL4 Mouse Monoclonal Antibody [clone: B-G28]
Antibodies
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: B-G28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR