Anti-IL-12 P35/p70 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-T21]

Supplier: Cell Sciences
CDM272
102995-764EA 443.08 USD
102995-764
Anti-IL-12 P35/p70 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-T21]
Antibodies
Suitable for intra-cytoplasmic flow cytometry

Type: Primary
Antigen: IL-12 P35/p70
Clonality: Monoclonal
Clone: B-T21
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR