Anti-DCR1 Mouse Monoclonal Antibody [clone: B-D44]

Supplier: Cell Sciences
CDM238
102995-730EA 553.85 USD
102995-730
Anti-DCR1 Mouse Monoclonal Antibody [clone: B-D44]
Antibodies
Inhibits TRAIL binding to TRAIL R3

Type: Primary
Antigen: DCR1
Clonality: Monoclonal
Clone: B-D44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR