Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DR5 Mouse Monoclonal Antibody [clone: B-D37]

Supplier: Cell Sciences
CDM237
102995-728
Anti-DR5 Mouse Monoclonal Antibody [clone: B-D37]
Agonist: induced apoptosis on Jurkat cells. Enhances TRAIL induced apoptosis of U937 cell line.

Type: Primary
Antigen: DR5
Clonality: Monoclonal
Clone: B-D37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty