Anti-DR4 Mouse Monoclonal Antibody [clone: B-N36]

Supplier: Cell Sciences
CDM231
102995-716EA 553.85 USD
102995-716
Anti-DR4 Mouse Monoclonal Antibody [clone: B-N36]
Antibodies
Neutralizes TRAIL induced apoptosis on HEL cell line

Type: Primary
Antigen: DR4
Clonality: Monoclonal
Clone: B-N36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR