Anti-TEK Mouse Monoclonal Antibody [clone: B-B48]

Supplier: Cell Sciences
CDM230
102995-714EA 443.08 USD
102995-714
Anti-TEK Mouse Monoclonal Antibody [clone: B-B48]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: TEK
Clonality: Monoclonal
Clone: B-B48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR