Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD58 Mouse Monoclonal Antibody [clone: B-L28]

Supplier: Cell Sciences
CDM227
102995-708
Anti-CD58 Mouse Monoclonal Antibody [clone: B-L28]
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: B-L28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty