Anti-CD58 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-L28]

Supplier: Cell Sciences
CDM226
102995-706EA 609.23 USD
102995-706
Anti-CD58 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-L28]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: B-L28
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR