Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD34 Mouse Monoclonal Antibody [clone: B-C34]

Supplier: Cell Sciences
CDM218
102995-690EA 418.46 USD InStock
102995-690
Anti-CD34 Mouse Monoclonal Antibody [clone: B-C34]
Antibodies
Immunohistochemistry: Recommended dilution 1:50

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: B-C34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty