Anti-CD151 Mouse Monoclonal Antibody [clone: B-E31]

Supplier: Cell Sciences
CDM207
102995-674EA 553.85 USD
102995-674
Anti-CD151 Mouse Monoclonal Antibody [clone: B-E31]
Antibodies
Type: Primary
Antigen: CD151
Clonality: Monoclonal
Clone: B-E31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR