Anti-CD86 Mouse Monoclonal Antibody [clone: B-T7]

Supplier: Cell Sciences
CDM201
102995-670EA 553.85 USD
102995-670
Anti-CD86 Mouse Monoclonal Antibody [clone: B-T7]
Antibodies
Inhibits Mixed Lymphocyte Culture

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: B-T7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR