Anti-CD75 Mouse Monoclonal Antibody [clone: B-L5]

Supplier: Cell Sciences
CDM197
102995-668EA 443.08 USD
102995-668
Anti-CD75 Mouse Monoclonal Antibody [clone: B-L5]
Antibodies
Type: Primary
Antigen: CD75
Clonality: Monoclonal
Clone: B-L5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR