Anti-PTPRC Mouse Monoclonal Antibody [clone: B-A11]

Supplier: Cell Sciences
CDM188
102995-650EA 443.08 USD
102995-650
Anti-PTPRC Mouse Monoclonal Antibody [clone: B-A11]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: B-A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR