Anti-PTPRC Mouse Monoclonal Antibody [clone: B-A11]

Supplier: Cell Sciences
CDM187
102995-648EA 553.85 USD
102995-648
Anti-PTPRC Mouse Monoclonal Antibody [clone: B-A11]
Antibodies
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: B-A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR