Anti-CD40L Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-B29]

Supplier: Cell Sciences
CDM184
102995-642EA 443.08 USD
102995-642
Anti-CD40L Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-B29]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: B-B29
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR