Anti-CD3 Mouse Monoclonal Antibody (Biotin) [clone: B-B11]

Supplier: Cell Sciences
CDM129
102995-630EA 498.46 USD
102995-630
Anti-CD3 Mouse Monoclonal Antibody (Biotin) [clone: B-B11]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: B-B11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR