Anti-CD1b/CD1c Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-B5]

Supplier: Cell Sciences
CDM125
102995-622EA 443.08 USD
102995-622
Anti-CD1b/CD1c Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-B5]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD1b/CD1c
Clonality: Monoclonal
Clone: B-B5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR