Anti-CD1b/CD1c Mouse Monoclonal Antibody [clone: B-B5]

Supplier: Cell Sciences
CDM124
102995-620EA 553.85 USD
102995-620
Anti-CD1b/CD1c Mouse Monoclonal Antibody [clone: B-B5]
Antibodies
Type: Primary
Antigen: CD1b/CD1c
Clonality: Monoclonal
Clone: B-B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR