Anti-IL17A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-A50]

Supplier: Cell Sciences
CDM117
102995-606EA 443.08 USD
102995-606
Anti-IL17A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-A50]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: B-A50
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR