Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL23A Mouse Monoclonal Antibody [clone: B-Z23]

Supplier: Cell Sciences
CDM115
102995-602EA 789.23 USD InStock
102995-602
Anti-IL23A Mouse Monoclonal Antibody [clone: B-Z23]
Antibodies
Neutralizes the bioactivity of rhIL-23 on mouse splenocytes C57BI/6 (measurement of mIL-17 or mIL-22 production). Suitable for immunochemistry on paraffin embedded section.

Type: Primary
Antigen: IL23A
Clonality: Monoclonal
Clone: B-Z23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty