Anti-DR4 Mouse Monoclonal Antibody [clone: B-T35]

Supplier: Cell Sciences
CDM113
102995-598EA 553.85 USD
102995-598
Anti-DR4 Mouse Monoclonal Antibody [clone: B-T35]
Antibodies
Type: Primary
Antigen: DR4
Clonality: Monoclonal
Clone: B-T35
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR