Anti-IL10 Mouse Monoclonal Antibody [clone: B-T10]

Supplier: Cell Sciences
CDM111
102995-594EA 955.38 USD
102995-594
Anti-IL10 Mouse Monoclonal Antibody [clone: B-T10]
Antibodies
Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: B-T10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR