Anti-TM Mouse Monoclonal Antibody [clone: B-S24]

Supplier: Cell Sciences
CDM109
102995-592EA 553.85 USD
102995-592
Anti-TM Mouse Monoclonal Antibody [clone: B-S24]
Antibodies
Type: Primary
Antigen: TM
Clonality: Monoclonal
Clone: B-S24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR