Anti-IL7 Mouse Monoclonal Antibody [clone: B-S16]

Supplier: Cell Sciences
CDM108
102995-590EA 955.38 USD
102995-590
Anti-IL7 Mouse Monoclonal Antibody [clone: B-S16]
Antibodies
Recognizes the soluble antigen

Type: Primary
Antigen: IL7
Clonality: Monoclonal
Clone: B-S16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR