Anti-P53 Mouse Monoclonal Antibody [clone: B-F34]

Supplier: Cell Sciences
CDM091
102995-574EA 553.85 USD
102995-574
Anti-P53 Mouse Monoclonal Antibody [clone: B-F34]
Antibodies
Type: Primary
Antigen: P53
Clonality: Monoclonal
Clone: B-F34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR