Anti-SDC1 Mouse Monoclonal Antibody [clone: B-D30]

Supplier: Cell Sciences
CDM089
102995-570EA 553.85 USD
102995-570
Anti-SDC1 Mouse Monoclonal Antibody [clone: B-D30]
Antibodies
Type: Primary
Antigen: SDC1
Clonality: Monoclonal
Clone: B-D30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR