Anti-CR2 Mouse Monoclonal Antibody [clone: B-C6]

Supplier: Cell Sciences
CDM087
102995-566EA 553.85 USD
102995-566
Anti-CR2 Mouse Monoclonal Antibody [clone: B-C6]
Antibodies
Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: B-C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR