Anti-CXCL10/IP-10 Mouse Monoclonal Antibody [clone: B-C50]

Supplier: Cell Sciences
CDM085
102995-562EA 443.08 USD
102995-562
Anti-CXCL10/IP-10 Mouse Monoclonal Antibody [clone: B-C50]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CXCL10/IP-10
Clonality: Monoclonal
Clone: B-C50
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR