Anti-TRAIL Mouse Monoclonal Antibody [clone: B-S23]

Supplier: Cell Sciences
CDM078
102995-550EA 443.08 USD
102995-550
Anti-TRAIL Mouse Monoclonal Antibody [clone: B-S23]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: TRAIL
Clonality: Monoclonal
Clone: B-S23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR